Assay Details
| Gene Symbol: REN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341680.1 | NP_001320057.1 | 11 - 12 | 1320 | 69 | |
| NM_001341681.1 | NP_001329908.1 | 12 - 13 | 1205 | 69 | ||
| NM_001341682.1 | NP_001329909.1 | 12 - 13 | 977 | 69 | ||
More Information
Set Membership: |
|

REN1
Genomic Map