Assay Details
| Gene Symbol: REN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341680.1 | NP_001320057.1 | 3 - 4 | 626 | 91 | |
| NM_001341681.1 | NP_001329908.1 | 4 - 5 | 511 | 91 | ||
| NM_001341682.1 | NP_001329909.1 | 4 - 5 | 283 | 91 | ||
More Information
Set Membership: |
|

REN1
Genomic Map