Assay Details
| Gene Symbol: REN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341680.1 | NP_001320057.1 | 9 - 10 | 1186 | 76 | |
| NM_001341681.1 | NP_001329908.1 | 10 - 11 | 1071 | 76 | ||
| NM_001341682.1 | NP_001329909.1 | 10 - 11 | 843 | 76 | ||
More Information
Set Membership: |
|

REN1
Genomic Map