Assay Details
| Gene Symbol: PGI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084975.1 | NP_001078444.1 | 8 - 9 | 1427 | 95 | |
| NM_118595.5 | NP_194193.2 | 8 - 9 | 1427 | 95 | ||
| GenBank mRNA | AK227111.1 | - | 8 - 9 | 1265 | 95 | |
| BT000953.1 | - | 8 - 9 | 1212 | 95 | ||
More Information
Set Membership: |
|

PGI1
Genomic Map