Assay Details
| Gene Symbol: FC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341699.1 | NP_001329107.1 | 1 - 2 | 279 | 67 | |
| NM_001341700.1 | NP_001329103.1 | 1 - 2 | 279 | 67 | ||
| NM_001341705.1 | NP_001329109.1 | 1 - 2 | 235 | 67 | ||
| NM_118607.3 | NP_194205.1 | 1 - 2 | 313 | 67 | ||
| GenBank mRNA | AK227055.1 | - | 1 - 2 | 185 | 67 | |
| BT025991.1 | - | 1 - 2 | 100 | 67 | ||
| D45353.1 | - | 1 - 2 | 220 | 67 | ||
| U16177.1 | - | 1 - 2 | 143 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001341705.1 transcripts.
Set Membership: |
|

FC2
Genomic Map