Assay Details
| Gene Symbol: AMY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118632.3 | NP_567714.1 | 1 - 2 | 111 | 99 | |
| GenBank mRNA | AY065233.1 | - | 1 - 2 | 111 | 99 | |
| AY117294.1 | - | 1 - 2 | 91 | 99 | ||
More Information
Set Membership: |
|

AMY1
Genomic Map