Assay Details
| Gene Symbol: AMY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118632.3 | NP_567714.1 | 3 - 4 | 675 | 139 | |
| GenBank mRNA | AY065233.1 | - | 3 - 4 | 675 | 139 | |
| AY117294.1 | - | 3 - 4 | 655 | 139 | ||
More Information
Set Membership: |
|

AMY1
Genomic Map