Assay Details
| Gene Symbol: AT4G25370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118669.3 | NP_567718.1 | 1 - 2 | 280 | 74 | |
| GenBank mRNA | AY035135.1 | - | 1 - 2 | 257 | 74 | |
| AY059070.1 | - | 1 - 2 | 218 | 74 | ||
| AY086824.1 | - | 1 - 2 | 249 | 74 | ||
More Information
Set Membership: |
|

AT4G25370
Genomic Map