Assay Details
| Gene Symbol: AT4G25370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118669.3 | NP_567718.1 | 2 - 3 | 321 | 75 | |
| GenBank mRNA | AY035135.1 | - | 2 - 3 | 298 | 75 | |
| AY059070.1 | - | 2 - 3 | 259 | 75 | ||
| AY086824.1 | - | 2 - 3 | 290 | 75 | ||
More Information
Set Membership: |
|

AT4G25370
Genomic Map