Assay Details
| Gene Symbol: AT4G25370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118669.3 | NP_567718.1 | 4 - 5 | 609 | 63 | |
| GenBank mRNA | AY035135.1 | - | 4 - 5 | 586 | 63 | |
| AY059070.1 | - | 4 - 5 | 547 | 63 | ||
| AY086824.1 | - | 4 - 5 | 578 | 63 | ||
More Information
Set Membership: |
|

AT4G25370
Genomic Map