Assay Details
| Gene Symbol: RS40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084978.3 | NP_001078447.1 | 6 - 7 | 1460 | 52 | |
| NM_001203908.1 | NP_001190837.1 | 6 - 7 | 1170 | 52 | ||
| NM_001341745.1 | NP_001320063.1 | 5 - 6 | 1659 | 52 | ||
| NM_001341746.1 | NP_001328585.1 | 4 - 5 | 1985 | 52 | ||
| NM_001341747.1 | NP_001328587.1 | 5 - 6 | 1762 | 52 | ||
| NM_001341748.1 | NP_001328586.1 | 4 - 5 | 1687 | 52 | ||
| NM_118682.4 | NP_194280.1 | 5 - 6 | 1203 | 52 | ||
| NM_202887.3 | NP_974616.2 | 3 - 4 | 1134 | 52 | ||
| GenBank mRNA | AK317023.1 | - | 6 - 7 | 1384 | 52 | |
| X99437.1 | - | 1163 | 52 | |||
More Information
Set Membership: |
|

RS40
Genomic Map