Assay Details
| Gene Symbol: SLK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341749.1 | NP_001320064.1 | 1 - 2 | 1284 | 87 | |
| NM_118684.9 | NP_194282.2 | 2 - 3 | 1077 | 87 | ||
| GenBank mRNA | AK226715.1 | - | 2 - 3 | 825 | 87 | |
More Information
Set Membership: |
|

SLK1
Genomic Map