Assay Details
| Gene Symbol: ACD1-LIKE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118697.4 | NP_567725.1 | 1 - 2 | 626 | 75 | |
| NM_179113.2 | NP_849444.1 | 1 - 2 | 563 | 75 | ||
| GenBank mRNA | AF424575.1 | - | 1 - 2 | 532 | 75 | |
| AY056168.1 | - | 1 - 2 | 555 | 75 | ||
| AY062731.1 | - | 1 - 2 | 563 | 75 | ||
| AY344062.1 | - | 1 - 2 | 519 | 75 | ||
| BT002586.1 | - | 1 - 2 | 519 | 75 | ||
More Information
Set Membership: |
|

ACD1-LIKE
Genomic Map