Assay Details
| Gene Symbol: ACD1-LIKE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118697.4 | NP_567725.1 | 4 - 5 | 1078 | 105 | |
| NM_179113.2 | NP_849444.1 | 3 - 4 | 1084 | 105 | ||
| GenBank mRNA | AF424575.1 | - | 4 - 5 | 984 | 105 | |
| AY056168.1 | - | 4 - 5 | 1007 | 105 | ||
| AY062731.1 | - | 3 - 4 | 1084 | 105 | ||
| AY344062.1 | - | 4 - 5 | 971 | 105 | ||
| BT002586.1 | - | 3 - 4 | 1040 | 105 | ||
More Information
Set Membership: |
|

ACD1-LIKE
Genomic Map