Assay Details
| Gene Symbol: BETA-OHASE 1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118702.4 | NP_194300.1 | 4 - 5 | 832 | 94 | |
| GenBank mRNA | AF125576.1 | - | 4 - 5 | 718 | 94 | |
| AF370220.1 | - | 4 - 5 | 712 | 94 | ||
| AY113923.1 | - | 4 - 5 | 649 | 94 | ||
More Information
Set Membership: |
|

BETA-OHASE 1
Genomic Map