Assay Details
| Gene Symbol: BETA-OHASE 1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341759.1 | NP_001320065.1 | 5 - 6 | 1381 | 132 | |
| NM_118702.4 | NP_194300.1 | 6 - 7 | 1007 | 132 | ||
| GenBank mRNA | AF125576.1 | - | 6 - 7 | 893 | 132 | |
| AF370220.1 | - | 6 - 7 | 887 | 132 | ||
| AY113923.1 | - | 6 - 7 | 824 | 132 | ||
More Information
Set Membership: |
|

BETA-OHASE 1
Genomic Map