Assay Details
| Gene Symbol: QC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036639.3 | NP_001031716.1 | 3 - 4 | 494 | 127 | |
| NM_001036640.1 | NP_001031717.1 | 3 - 4 | 478 | 127 | ||
| NM_001341762.1 | NP_001329256.1 | 3 - 4 | 458 | 127 | ||
| NM_001341763.1 | NP_001329255.1 | 3 - 4 | 492 | 127 | ||
| NM_118704.2 | NP_567727.1 | 3 - 4 | 494 | 127 | ||
| GenBank mRNA | AK316976.1 | - | 3 - 4 | 427 | 127 | |
| AK318678.1 | - | 3 - 4 | 480 | 127 | ||
| BT001929.1 | - | 3 - 4 | 370 | 127 | ||
More Information
Set Membership: |
|

QC
Genomic Map