Assay Details
| Gene Symbol: QC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036639.3 | NP_001031716.1 | 8 - 9 | 907 | 63 | |
| NM_001036640.1 | NP_001031717.1 | 8 - 9 | 831 | 63 | ||
| NM_118704.2 | NP_567727.1 | 8 - 9 | 907 | 63 | ||
| GenBank mRNA | AK316976.1 | - | 8 - 9 | 840 | 63 | |
| AK318678.1 | - | 7 - 8 | 1191 | 63 | ||
More Information
Set Membership: |
|

QC
Genomic Map