Assay Details
| Gene Symbol: BGAL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341777.1 | NP_001329560.1 | 10 - 11 | 1250 | 79 | |
| NM_001341778.1 | NP_001329554.1 | 10 - 11 | 1192 | 79 | ||
| NM_001341779.1 | NP_001329557.1 | 10 - 11 | 1192 | 79 | ||
| NM_001341780.1 | NP_001329558.1 | 10 - 11 | 1193 | 79 | ||
| NM_001341781.1 | NP_001329559.1 | 10 - 11 | 1193 | 79 | ||
| NM_001341782.1 | NP_001329555.1 | 10 - 11 | 1192 | 79 | ||
| NM_001341783.1 | NP_001329556.1 | 10 - 11 | 1192 | 79 | ||
| NM_118747.3 | NP_194344.2 | 10 - 11 | 1250 | 79 | ||
| NM_179222.2 | NP_849553.1 | 10 - 11 | 1479 | 79 | ||
| GenBank mRNA | AJ270308.1 | - | 10 - 11 | 1089 | 79 | |
| AY099612.1 | - | 10 - 11 | 1192 | 79 | ||
| BT000267.1 | - | 10 - 11 | 1089 | 79 | ||
More Information
Set Membership: |
|

BGAL12
Genomic Map