Assay Details
| Gene Symbol: BGAL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341780.1 | NP_001329558.1 | 16 - 17 | 2286 | 129 | |
| NM_001341781.1 | NP_001329559.1 | 16 - 17 | 2255 | 129 | ||
| NM_118747.3 | NP_194344.2 | 16 - 17 | 2337 | 129 | ||
| NM_179222.2 | NP_849553.1 | 17 - 18 | 2466 | 129 | ||
| GenBank mRNA | AJ270308.1 | - | 17 - 18 | 2076 | 129 | |
| AY099612.1 | - | 16 - 17 | 2279 | 129 | ||
More Information
Set Membership: |
|

BGAL12
Genomic Map