Assay Details
| Gene Symbol: MIOX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203915.2 | NP_001190844.1 | 7 - 8 | 838 | 106 | |
| NM_118759.5 | NP_194356.2 | 7 - 8 | 943 | 106 | ||
| GenBank mRNA | AY142501.1 | - | 7 - 8 | 699 | 106 | |
More Information
Set Membership: |
|

MIOX4
Genomic Map