Assay Details
| Gene Symbol: emb1027 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341791.1 | NP_001329500.1 | 8 - 9 | 996 | 80 | |
| NM_001341792.1 | NP_001329501.1 | 8 - 9 | 992 | 80 | ||
| NM_001341793.1 | NP_001329502.1 | 8 - 9 | 906 | 80 | ||
| NM_001341794.1 | NP_001329503.1 | 8 - 9 | 902 | 80 | ||
| NM_118763.3 | NP_194360.1 | 8 - 9 | 858 | 80 | ||
More Information
Set Membership: |
|

emb1027
Genomic Map