Assay Details
| Gene Symbol: AT4G26630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036647.3 | NP_001031724.1 | 9 - 10 | 2173 | 66 | |
| NM_118797.5 | NP_194393.3 | 9 - 10 | 2164 | 66 | ||
More Information
Set Membership: |
|

AT4G26630
Genomic Map