Assay Details
| Gene Symbol: EXT-like | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118809.5 | NP_194405.1 | 1 - 2 | 236 | 73 | |
| GenBank mRNA | AY048219.1 | - | 1 - 2 | 150 | 73 | |
| AY094009.1 | - | 1 - 2 | 95 | 73 | ||
More Information
Set Membership: |
|

EXT-like
Genomic Map