Assay Details
| Gene Symbol: AR192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118812.4 | NP_567757.1 | 1 - 2 | 442 | 75 | |
| GenBank mRNA | AK118205.1 | - | 1 - 2 | 282 | 75 | |
| AY087425.1 | - | 1 - 2 | 259 | 75 | ||
| BT006047.1 | - | 1 - 2 | 144 | 75 | ||
More Information
Set Membership: |
|

AR192
Genomic Map