Assay Details
| Gene Symbol: EMB2788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341833.1 | NP_001320074.1 | 4 - 5 | 2516 | 94 | |
| NM_118835.3 | NP_194431.3 | 4 - 5 | 2516 | 94 | ||
More Information
Set Membership: |
|

EMB2788
Genomic Map