Assay Details
| Gene Symbol: EMB2788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118835.3 | NP_194431.3 | 7 - 8 | 6165 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMB2788| Gene Symbol: EMB2788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118835.3 | NP_194431.3 | 7 - 8 | 6165 | 86 | |
Set Membership: |
|