Assay Details
| Gene Symbol: EMB2788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341833.1 | NP_001320074.1 | 10 - 11 | 6454 | 114 | |
| NM_118835.3 | NP_194431.3 | 8 - 9 | 6370 | 114 | ||
More Information
Set Membership: |
|

EMB2788
Genomic Map