Assay Details
| Gene Symbol: VPS22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125593.1 | NP_001119065.1 | 4 - 5 | 845 | 105 | |
| NM_001125594.1 | NP_001119066.1 | 4 - 5 | 658 | 105 | ||
| NM_001125595.2 | NP_001119067.1 | 6 - 7 | 761 | 105 | ||
| NM_001341834.1 | NP_001328604.1 | 4 - 5 | 1023 | 105 | ||
| NM_001341835.1 | NP_001328603.1 | 4 - 5 | 1003 | 105 | ||
| NM_118838.5 | NP_194434.2 | 6 - 7 | 812 | 105 | ||
More Information
Set Membership: |
|

VPS22
Genomic Map