Assay Details
| Gene Symbol: VPS22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125593.1 | NP_001119065.1 | 5 - 6 | 992 | 98 | |
| NM_001125594.1 | NP_001119066.1 | 5 - 6 | 805 | 98 | ||
| NM_001125595.2 | NP_001119067.1 | 7 - 8 | 908 | 98 | ||
| NM_001341834.1 | NP_001328604.1 | 5 - 6 | 1170 | 98 | ||
| NM_001341835.1 | NP_001328603.1 | 5 - 6 | 1150 | 98 | ||
| NM_118838.5 | NP_194434.2 | 7 - 8 | 959 | 98 | ||
More Information
Set Membership: |
|

VPS22
Genomic Map