Assay Details
| Gene Symbol: VPS22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125593.1 | NP_001119065.1 | 7 - 8 | 1252 | 84 | |
| NM_001125594.1 | NP_001119066.1 | 7 - 8 | 1065 | 84 | ||
| NM_001125595.2 | NP_001119067.1 | 9 - 10 | 1168 | 84 | ||
| NM_001341834.1 | NP_001328604.1 | 7 - 8 | 1430 | 84 | ||
| NM_001341835.1 | NP_001328603.1 | 7 - 8 | 1410 | 84 | ||
| NM_118838.5 | NP_194434.2 | 9 - 10 | 1219 | 84 | ||
More Information
Set Membership: |
|

VPS22
Genomic Map