Assay Details
Gene Symbol: COBL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118845.3 | NP_567766.1 | 3 - 4 | 1113 | 99 | |
GenBank mRNA | AK175703.1 | - | 3 - 4 | 1079 | 99 | |
AK176063.1 | - | 3 - 4 | 1079 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
