Assay Details
| Gene Symbol: CIP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125596.2 | NP_001119068.1 | 4 - 5 | 550 | 77 | |
| NM_118877.4 | NP_194473.1 | 4 - 5 | 1222 | 77 | ||
| GenBank mRNA | AB012912.1 | - | 4 - 5 | 550 | 77 | |
More Information
Set Membership: |
|

CIP7
Genomic Map