Assay Details
| Gene Symbol: CIP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125596.2 | NP_001119068.1 | 8 - 9 | 2420 | 107 | |
| NM_118877.4 | NP_194473.1 | 8 - 9 | 3092 | 107 | ||
| GenBank mRNA | AB012912.1 | - | 8 - 9 | 2420 | 107 | |
More Information
Set Membership: |
|

CIP7
Genomic Map