Assay Details
| Gene Symbol: PORB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036654.1 | NP_001031731.1 | 1 - 2 | 260 | 86 | |
| NM_118879.4 | NP_194474.1 | 1 - 2 | 260 | 86 | ||
| GenBank mRNA | AK317009.1 | - | 1 - 2 | 215 | 86 | |
| AY042883.1 | - | 1 - 2 | 211 | 86 | ||
| AY054206.1 | - | 1 - 2 | 215 | 86 | ||
| AY081465.1 | - | 1 - 2 | 158 | 86 | ||
| U29785.1 | - | 1 - 2 | 158 | 86 | ||
More Information
Set Membership: |
|

PORB
Genomic Map