Assay Details
| Gene Symbol: CBSX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118881.3 | NP_194476.2 | 1 - 2 | 432 | 86 | |
| GenBank mRNA | BT026427.1 | - | 1 - 2 | 325 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CBSX5| Gene Symbol: CBSX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118881.3 | NP_194476.2 | 1 - 2 | 432 | 86 | |
| GenBank mRNA | BT026427.1 | - | 1 - 2 | 325 | 86 | |
Set Membership: |
|