Assay Details
| Gene Symbol: TPS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341861.1 | NP_001327998.1 | 4 - 5 | 752 | 141 | |
| NM_118890.2 | NP_194485.1 | 4 - 5 | 752 | 141 | ||
| GenBank mRNA | KJ676985.1 | - | 4 - 5 | 712 | 141 | |
More Information
Set Membership: |
|

TPS4
Genomic Map