Assay Details
| Gene Symbol: SPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338161.1 | NP_001327623.1 | 2 - 3 | 1496 | 88 | |
| NM_001338162.1 | NP_001327624.1 | 2 - 3 | 1496 | 88 | ||
| NM_001338163.1 | NP_001327621.1 | 2 - 3 | 1422 | 88 | ||
| NM_001338164.1 | NP_001327622.1 | 1 - 2 | 194 | 88 | ||
| NM_148725.4 | NP_683567.1 | 2 - 3 | 1428 | 88 | ||
| GenBank mRNA | HQ228994.1 | - | 2 - 3 | 871 | 88 | |
| HQ228995.1 | - | 2 - 3 | 871 | 88 | ||
More Information
Set Membership: |
|

SPA3
Genomic Map