Assay Details
Gene Symbol: SPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338161.1 | NP_001327623.1 | 3 - 4 | 2449 | 85 | |
NM_001338162.1 | NP_001327624.1 | 3 - 4 | 2449 | 85 | ||
NM_001338163.1 | NP_001327621.1 | 3 - 4 | 2375 | 85 | ||
NM_001338164.1 | NP_001327622.1 | 2 - 3 | 1147 | 85 | ||
NM_148725.4 | NP_683567.1 | 3 - 4 | 2381 | 85 | ||
GenBank mRNA | HQ228994.1 | - | 3 - 4 | 1824 | 85 | |
HQ228995.1 | - | 3 - 4 | 1824 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
