Assay Details
| Gene Symbol: AT3G32904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148784.1 | NP_683626.1 | 1 - 2 | 234 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G32904| Gene Symbol: AT3G32904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148784.1 | NP_683626.1 | 1 - 2 | 234 | 118 | |
Set Membership: |
|