Assay Details
| Gene Symbol: AT3G32904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148784.1 | NP_683626.1 | 2 - 3 | 373 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G32904| Gene Symbol: AT3G32904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148784.1 | NP_683626.1 | 2 - 3 | 373 | 83 | |
Set Membership: |
|