Assay Details
| Gene Symbol: EMB2423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339371.1 | NP_001325844.1 | 10 - 11 | 1956 | 79 | |
| NM_148864.4 | NP_680117.3 | 10 - 11 | 1944 | 79 | ||
More Information
Set Membership: |
|

EMB2423
Genomic Map