Assay Details
| Gene Symbol: BB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035845.2 | NP_001030922.1 | 3 - 4 | 678 | 65 | |
| NM_148885.3 | NP_680148.1 | 3 - 4 | 402 | 65 | ||
| GenBank mRNA | AK227673.1 | - | 3 - 4 | 399 | 65 | |
| BT010197.1 | - | 1 - 2 | 149 | 65 | ||
More Information
Set Membership: |
|

BB
Genomic Map