Assay Details
| Gene Symbol: ATFP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112005.4 | NP_187779.1 | 3 - 4 | 352 | 83 | |
| GenBank mRNA | AK118150.1 | - | 3 - 4 | 239 | 83 | |
| BT005576.1 | - | 2 - 3 | 132 | 83 | ||
| U64911.1 | - | 3 - 4 | 215 | 83 | ||
More Information
Set Membership: |
|

ATFP8
Genomic Map