Assay Details
| Gene Symbol: FOLB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112008.2 | NP_187781.1 | 1 - 2 | 167 | 86 | |
| GenBank mRNA | BT030075.1 | - | 1 - 2 | 53 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FOLB1| Gene Symbol: FOLB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112008.2 | NP_187781.1 | 1 - 2 | 167 | 86 | |
| GenBank mRNA | BT030075.1 | - | 1 - 2 | 53 | 86 | |
Set Membership: |
|