Assay Details
| Gene Symbol: MS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112032.3 | NP_187805.1 | 5 - 6 | 911 | 82 | |
| GenBank mRNA | EU280150.1 | - | 4 - 5 | 835 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MS2| Gene Symbol: MS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112032.3 | NP_187805.1 | 5 - 6 | 911 | 82 | |
| GenBank mRNA | EU280150.1 | - | 4 - 5 | 835 | 82 | |
Set Membership: |
|