Assay Details
| Gene Symbol: MS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112032.3 | NP_187805.1 | 8 - 9 | 1764 | 131 | |
| GenBank mRNA | EU280150.1 | - | 7 - 8 | 1688 | 131 | |
More Information
Set Membership: |
|

MS2
Genomic Map