Assay Details
| Gene Symbol: RBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202939.1 | NP_001189868.1 | 10 - 11 | 2270 | 107 | |
| NM_112064.5 | NP_566417.3 | 10 - 11 | 2459 | 107 | ||
| GenBank mRNA | AF245395.1 | - | 10 - 11 | 2177 | 107 | |
| AY150438.1 | - | 9 - 10 | 1973 | 107 | ||
More Information
Set Membership: |
|

RBR1
Genomic Map