Assay Details
| Gene Symbol: RBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202939.1 | NP_001189868.1 | 16 - 17 | 3048 | 133 | |
| NM_112064.5 | NP_566417.3 | 16 - 17 | 3237 | 133 | ||
| GenBank mRNA | AF245395.1 | - | 16 - 17 | 2955 | 133 | |
| AY150438.1 | - | 15 - 16 | 2751 | 133 | ||
More Information
Set Membership: |
|

RBR1
Genomic Map