Assay Details
| Gene Symbol: RBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112064.5 | NP_566417.3 | 17 - 18 | 3357 | 73 | |
| GenBank mRNA | AF245395.1 | - | 17 - 18 | 3075 | 73 | |
| AY150438.1 | - | 16 - 17 | 2871 | 73 | ||
More Information
Set Membership: |
|

RBR1
Genomic Map